Restriction enzymes, BamHI
Fornecedor: Promega
R6021
R6025
PROMR60212500EA
75.6
EUR
PROMR60212500
PROMR602512KU
Restriction enzymes, BamHI
Reagentes de ácidos nucleicos
Restriction Enzymes
These BamHI restriction enzyme recognises and cut G˅GATC C and C CTAG^G at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq® Green master mix
- Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
- Source: Streptomyces albus G
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 300 mM KCl, 5 mM MgCl₂, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: Kit includes 1×1 ml buffer E 10X buffer, 1×150 µl bovine serum albumin (acetylated), 1×2500 units BamHI and 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...