Print…
Coupled Reticulocyte Lysate Systems, TnT®
Fornecedor: Promega
L4600 L4601 L4610 L4611 L4950 L5010 L5020
PROML46000001EA 1260 EUR
PROML46000001 PROML46010001 PROML46100001 PROML46110001 PROML49500001 PROML50100001 PROML50200001
Coupled Reticulocyte Lysate Systems, TnT®
Expressão de proteínas Protein Expression Systems
TnT® Coupled Reticulocyte Lysate System offers researchers an alternative for eukaryotic cell-free protein expression. It greatly simplifies the process and reduces the time required to obtain in vitro translation results.
  • Study protein:protein, protein:nucleic acid and protein expression interactions
  • Generate proteins in one hour versus days
Fornecimento: TnT® Rabbit Reticulocyte Lysate is supplied in 200 µl aliquots. Each system contains sufficient reagents to perform
approximately 40×50 µl translation reactions. Includes:
1 ml TnT® Rabbit Reticulocyte Lysate
90 µl TnT® Reaction Buffer
60 µl TnT® T3, T7 or SP6 RNA Polymerase (2×30 µl in dual systems)
5 µg Luciferase T3, T7 or SP6 Control DNA, 0,5 mg/ml
50 µl Amino Acid Mixture, minus Methionine, 1 mM
50 µl Amino Acid Mixture, minus Leucine, 1 mM
50 µl Amino Acid Mixture, minus Cysteine, 1 mM
250 µl Luciferase Assay Reagent
1× Luciferase Assay Wells (set of 3)
100 µl Magnesium Acetate (only with Cat. Nos. L4600 and L4601)