Print…
Competent cells, E. coli
Fornecedor: Promega
L1001 L1191 L2001 L2011 L1195 L2005
PROML10010001EA 184 EUR
PROML10010001 PROML11910001 PROML20010001 PROML20110001 PROML1195 PROML2005
Competent cells, E. coli
Células competentes
The E. coli competent cells, prepared according to a modified procedure of Hanahan, can be used for many standard molecular biology applications.
  • Cells are ready to use, no preparation time necessary
  • Transformation efficiencies guaranteed
  • No undesirable recombination events
JM109 competent cells are available for convenient transformation in two efficiencies: High Efficiency at greater than 108 cfu/µg and Subcloning Efficiency at greater than 107 cfu/µg. HB101 competent cells are available in high efficiency at greater than 108 cfu/µg. JM109 cells areMais informação do produto