Print…
CellTiter-Glo® 3D cell viability assay
Fornecedor: Promega
G9681 G9682 G9683
PROMG9681EA 99.8 EUR
PROMG9681 PROMG9682 PROMG9683
CellTiter-Glo® 3D cell viability assay
Ensaios Ensaios celulares
The CellTiter-Glo® 3D Cell Viability Assay is designed for determining cell viability in 3D microtissue spheroids. The assay reagent penetrates large spheroids and has increased lytic capacity allowing more accurate determination of viability compared to other assay methods.
  • Accurate 3D cytotoxicity determination
  • Simple, 30-minute protocol
  • Compatible with a variety of 3D culture methods
CellTiter-Glo® 3D Cell Viability Assay measures ATP as an indicator of viability and generates a luminescent readout that is much more sensitive than colorimetric or fluorescence-based methods.