Print…
Cytotoxicity assays CytoTox-Glo™
Fornecedor: Promega
G9291 G9292 G9290
G9291EA 796 EUR
G9291 PROMG9292 G9290
Cytotoxicity assays CytoTox-Glo™
Ensaios Ensaios celulares
The CytoTox-Glo™ Assay is a luminescent cytotoxicity assay that measures the relative number of dead cells in cell populations.
  • Measures extracellular activity of dead-cell protease
  • Easy and fast 'add-mix-measure' protocol
The CytoTox-Glo™ Assay measures the extracellular activity of a distinct intracellular protease activity (dead-cell protease) when the protease is released from membrane-compromised cells. A luminogenic cell-impermeant peptide substrate (AAF-aminoluciferin) is used to measure dead-cell protease activity and the liberated aminoluciferin product is measured as 'glow type' luminescence generated by the recombinant Luciferase provided in the assay reagent.
Fornecimento: Kit includes AAF-Glo™ Substrate, Assay Buffer and Digitonin.