MultiTox-Fluor multiplex cytotoxicity assay
Fornecedor: Promega
The MultiTox-Fluor Multiplex Cytotoxicity Assay is a single-reagent-addition, homogeneous, fluorescent assay measures cell viability and cytotoxicity by detecting two distinct protease activities. The live-cell protease activity is restricted to intact viable cells and is measured using a fluorogenic, cell-permeant peptide substrate (GF-AFC Substrate). A cell-impermeant, fluorogenic peptide substrate (bis-AAF-R110 substrate) is used to measure dead-cell protease activity that has been released from cells that have lost membrane integrity.
- Assay normalises data for cell number
- Reduces false-positive and false-negative results
- Multiplexing capability results in more data per well
- Complementary live and dead cell measures with independent chemistries serve as internal controls for each other
Fornecimento: The complete assay kit includes bis-AAF-R110 substrate, GF-AFC substrate and assay buffer.