SacI (from Streptomyces achromogenes)
Fornecedor: Promega
R6061
R6065
PROMR60611000EA
83
EUR
PROMR60611000
PROMR60655000
SacI (from Streptomyces achromogenes)
Enzimas
SacI recognises the same DNA sequence but cleaves at a different nucleotide than its neoschizomer, EcoICRI. SacI recognises G AGCT▾C and C▴TCGA G.
- Blue or white cloning qualified
- Higher level of quality control for enzymes used in cloning applications
- With incubation condition of buffer J. 37 °C
- Source: Streptomyces achromogenes
- Highly concentrated
Storage Buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA, 50% glycerol.
Fornecimento: Kit includes 1×1 ml buffer J 10X buffer, 1×150 µl bovine serum albumin, acetylated with 10 mg/ml concentration, 1×1,000 U SacI, 1×0,25 ml MULTI-CORE™ 10X buffer.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...