Print…
Probes and accessories for Vibra-Cell™ ultrasonic processors
Fornecedor: SONICS MATERIALS
630-0310 630-0409 630-0418 630-0731 630-0444 830-00004 630-0207 630-0208 630-0210 630-0209 630-0421 830-00118 830-00116 630-0423 630-0435 630-0419 830-00060 630-0220 630-0217 630-0218 630-0306 630-0420 630-0219 630-0559 630-0579 630-0422 630-0580 630-0608 630-0406 630-0651 630-0407 830-00130
SONI630-0310EA 1220 EUR
SONI630-0310 432-0111 432-0112 432-0117 432-0119 432-0128 432-0102 432-0103 432-0105 432-0104 432-0115 432-0131 432-0130 432-0116 432-0118 432-0113 432-0129 432-0107 SONI630-0217 SONI630-0218 SONI630-0306 432-0114 432-0106 432-0123 432-0125 432-0127 SONI630-0580 SONI630-0608 432-0267 432-0269 SONI630-0407 SONI830-00130
Probes and accessories for Vibra-Cell™ ultrasonic processors
Homogeneizadores Sonicadores
A wide range of probes for a variety of applications is available, including stepped and tapered microtips and also probes for processing in microplates.
  • Multi-element probes for deep well microplates
  • Cuphorns for non invasive sonication of samples
  • Continouous flow cells for dispersing and homogenising large volumes of low viscosity samples where extended exposure to ultrasonics is not desirable
  • Footswitch for hands-free operation
  • Sound abating enclosures - reduce noise and act as a containment facility
Probes for processing in microplates:
The multi-element probes are specifically designed for processing up to 96 deep well plates as well as 1,5/2,0 ml microtubes and 10 ml test tubes. Although originally designed for genomic and proteomic applications, these probes can also be used for cell and tissue fractionation, toxicity studies, sample preparations, ELISAs, enzymology, protein purification for RNA, DNA, PCR labelling, hybridisation, receptor binding studies, preparation of liposomes, micro emulsions, extractions, desegregation and dissolution.
Unlike ultrasonic baths, or microplate horns, the probes deliver the intensity directly into the sample, not through the microplate walls and unlike single probes with grooved protrusions, these composite probes act as mechanical amplifiers. These 20 kHz multi-element probes consist of a coupler and multiple 3 mm replaceable stepped microtips, spaced 18 mm apart. The complete assembly is approximately 260 mm long.
The probes can be mounted onto a heavy duty laboratory stand or incorporated into an automated X - Y positioning system to satisfy high throughput requirements.
Informações de encomenda: For details of the full range of acccesories including full wave probes, high gain probes, full wave extenders plus accessories for model VCX 1500, please contact your local Avantor office.
Atenção: *Safety Note: Never use a probe with replacement tip with low surface tension liquids (e.g. solvents). Use a solid probe instead.
** Can also be used with the VCX130 and VCX130PB