Print…

DNA (Deoxyribonucleic acid) (de esperma de arenque)

Fornecedor: Promega

D1811 D1815 D1816
PROMD18110010EA 108 EUR
PROMD18110010 PROMD18150100 PROMD18160500
DNA (Deoxyribonucleic acid) (de esperma de arenque)
DNA (Deoxyribonucleic acid)

Herring sperm DNA is useful as a blocking agent in nucleic acid hybridisation experiments.


  • Quality tested and certified free of DNase or RNase activity
  • Provided as a ready-to-use 10 mg/ml solution
  • Nucleic acid molecular weight marker


Storage buffer: 10 mM Tris-HCl (pH 7,5), 10 mM NaCl, 1 mM EDTA.


Atenção: Product may be viscous at 4 °C. Prior to use, ensure product is at room temperature (it may be briefly warmed at 37 °C) and mixed thoroughly to ensure homogeneity.

Número MDL : MFCD00130922
Número CAS: 9007-49-2

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR