Print…
Streptavidin, paramagnetic particles, MagneSphere®
Fornecedor: Promega
Z5481 Z5482
PROMZ54810009EA 268 EUR
PROMZ54810009 PROMZ54820025
Streptavidin, paramagnetic particles, MagneSphere®
Esferas Esferas magnéticas
The streptavidin MagneSphere® paramagnetic particles (PMPs) may be used in the magnetic separation and purification of a wide variety of biotinylated nucleic acid or protein molecules. The particles are quality tested and approved for isolation of biotinylated nucleic acids, proteins and antibodies.