Print…
Magnetic separation stand, MagneSphere® technology
Fornecedor: Promega
Z5331 Z5332 Z5333 Z5341 Z5342 Z5343 Z5410
PROMZ53310500EA 200 EUR
PROMZ53310500 PROMZ53321500 PROMZ53330075 PROMZ53410500 PROMZ5342 PROMZ5343 PROMZ54100001
Magnetic separation stand, MagneSphere® technology
Suportes/Racks Suportes para separação magnética
The MagneSphere® Technology magnetic separation stands can be used in conjunction with any of the PolyATtract® systems and are ideal for applications requiring multiple paramagnetic isolations of biomolecules.
  • Use with both MagneSphere® and PolyATtract® System components
  • Conduct simultaneous processing of the contents of two tubes or twelve tubes of various sizes
These stands use the same strong rare earth magnet used in the PolyATtract® Systems Magnetic Separation Stands and come in a variety of sizes to accommodate 2 to 96 samples.