Protein purification system, MagneGST™
Fornecedor: Promega
The MagneGST™ protein purification system provides a simple, rapid and reliable method for the purification of glutathione-S-transferase (GST) fusion proteins. Immobilised glutathione paramagnetic particles are used to isolate GST-tagged protein directly from a crude or cleared E. coli lysate using either a manual or automated procedure.
- High-throughput purification of GST-tagged proteins using automated platforms
- High yields with little or no nonspecific background
GST-tagged proteins can be purified on a small scale from 1 ml of culture or on a large scale using more than 50 ml of culture. Samples also can be processed using a robotic platform.
Informações de encomenda: This product requires the use of a magnetic stand that is not included.
Fornecimento: System contains MagneGST™ binding/wash buffer (80 or 400 ml); Glutathione, 0,5 M (2 or 10 ml); MagneGST™ cell lysis reagent (12 or 60 ml); MagneGST™ glutathione particles (4 or 2×10 ml); RQ1 RNase-free DNase (1000 units).