CYP2C8 assay systems, P450-Glo™
Fornecedor: Promega
V8781
V8782
PROMV8781EA
243
EUR
PROMV8781
PROMV8782
CYP2C8 assay systems, P450-Glo™
Ensaios
Ensaios celulares
The P450-Glo CYP2C8 assays provide a homogeneous, luminescent method for measuring cytochrome CYP2C8 activity.
- Broad dynamic range, low background and better sensitivity for less ambiguous data
- Luminescent output eliminates interference from fluorescent test compounds
- Optimal data quality for microplate reader assay
- Simplified, homogeneous experimental protocol
Fornecimento: Assay system contains Luciferin-ME, 5 mM (1×300 or 2×750 µl); luciferin detection reagent (lyophilised) (1 vial); reconstitution buffer (1×10 or 1×50 ml).