Print…
CYP2C8 assay systems, P450-Glo™
Fornecedor: Promega
V8781 V8782
PROMV8781EA 243 EUR
PROMV8781 PROMV8782
CYP2C8 assay systems, P450-Glo™
Ensaios Ensaios celulares
The P450-Glo CYP2C8 assays provide a homogeneous, luminescent method for measuring cytochrome CYP2C8 activity.
  • Broad dynamic range, low background and better sensitivity for less ambiguous data
  • Luminescent output eliminates interference from fluorescent test compounds
  • Optimal data quality for microplate reader assay
  • Simplified, homogeneous experimental protocol
Fornecimento: Assay system contains Luciferin-ME, 5 mM (1×300 or 2×750 µl); luciferin detection reagent (lyophilised) (1 vial); reconstitution buffer (1×10 or 1×50 ml).