Print…
CYP1B1 assay systems, P450-Glo™
Fornecedor: Promega
V8761 V8762
PROMV8761EA 243 EUR
PROMV8761 PROMV8762
CYP1B1 assay systems, P450-Glo™
Ensaios Ensaios celulares
The P450-Glo CYP1B1 assays provide a homogeneous, luminescent method for measuring cytochrome CYP1B1 activity. These assays can be used to detect a mixture of CYP1A1 and CYP1B1 activities in cell types that express both enzymes.
  • Broad dynamic range, low background and better sensitivity for less ambiguous data
  • Luminescent output eliminates interference from fluorescent test compounds
  • Optimal data quality for microplate reader assay
  • Simplified, homogeneous experimental protocol
Fornecimento: Kit contains Luciferin-CEE, 5 mM (1×70 or 1×350 µl); luciferin detection reagent (lyophilised) (1 vial); reconstitution buffer (1×10 or 1×50 ml).