Restriction enzymes, Spe I
Fornecedor: Promega
R6591
R6595
PROMR65910200EA
124
EUR
PROMR65910200
PROMR65951000
Restriction enzymes, Spe I
Reagentes de ácidos nucleicos
Restriction Enzymes
These SpeI restriction enzyme recognises A˅CTAG T and T GATC^A sites and cuts best at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Blue and white cloning assay provides a higher level of quality control for enzymes used in cloning applications
- Restriction enzymes are assayed to ensure optimal performance in genomic analysis applications
- Ends generated with SpeI can be directly ligated to ends generated by NheI or XbaI
- Source: Sphaerotilus natans ATCC 13923
Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.
Fornecimento: Kit includes (1×200) (1×1000) U SpeI, (1×1 ml) buffer B 10X, (1×0,25 ml) MULTI-CORE™ 10X buffer, (1×150 µl) bovine serum albumin (acetylated).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...