Restriction enzymes, ClaI
Fornecedor: Promega
R6551
R6555
PROMR65510500EA
0
EUR
PROMR65510500
PROMR65552500
Restriction enzymes, ClaI
Reagentes de ácidos nucleicos
Restriction Enzymes
These ClaI restriction enzyme recognises AT˅CG AT and TA GC^TA sites and cuts best at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq® green master mix
- Blue and white cloning assay provides a higher level of quality control for enzymes used in cloning applications
- Restriction enzymes are assayed to ensure optimal performance in genomic analysis applications
- Sensitive to overlapping dam methylation
- Source: Caryophanon latum L
Storage buffer: 10 mM Tris-HCl (pH 7,4), 300 mM sodium glutamate, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.
Fornecimento: Kit includes (1×500) (1×2500) U ClaI, (1×1 ml) buffer E 10X, (1×0,25 ml) MULTI-CORE™ 10X buffer, (1×150 µl) bovine serum albumin (acetylated).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...