Restriction enzymes, NcoI
Fornecedor: Promega
R6513
R6515
PROMR65130200EA
124
EUR
PROMR65130200
PROMR65151000
Restriction enzymes, NcoI
Reagentes de ácidos nucleicos
Restriction Enzymes
These NcoI restriction enzyme recognises C˅CATG G and G GTAC^C sites and cuts best at 37 °C in its own unique buffer.
- Capable of digesting DNA in 15 minutes or less
- Active and capable of digestion directly in GoTaq® green master mix
- Blue and white cloning assay provides a higher level of quality control for enzymes used in cloning applications
- Source: Nocardia corallina
Storage buffer: 10 mM Tris-HCl (pH 7,4), 300 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.
Fornecimento: Kit includes 1×200 U NcoI, 1×1 ml buffer D 10X, 1×0,25 ml MULTI-CORE™ 10X buffer, 1×150 µl bovine serum albumin (acetylated).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...