Print…
Restriction enzymes, NheI
Fornecedor: Promega
R6501 R6505
PROMR65010250EA 108 EUR
PROMR65010250 PROMR65051250
Restriction enzymes, NheI
Reagentes de ácidos nucleicos Restriction Enzymes
These NheI restriction enzyme recognises G˅CTAG C and C GATC^G sites and cuts best at 37 °C in its own unique buffer.
  • Capable of digesting DNA in 15 minutes or less
  • Active and capable of digestion directly in GoTaq®green master mix
  • Restriction enzymes are assayed to ensure optimal performance in genomic analysis applications
  • Ends generated with NheI can be directly ligated to ends generated by SpeI or XbaI
  • Source: Neisseria mucosa heidelbergensis
Fornecimento: Kit includes (1×250) (1×1250U) NheI, (1×1 ml) buffer B 10X, (1×0,25 ml) MULTI-CORE™ 10X buffer, (1×150 µl) bovine serum albumin (acetylated).