Restriction enzymes, MspI
Fornecedor: Promega
R6401
R6405
PROMR64012000EA
0
EUR
PROMR64012000
PROMR640510KU
Restriction enzymes, MspI
Reagentes de ácidos nucleicos
Restriction Enzymes
These SspI restriction enzyme recognises C˅CG G and G GC^C sites and cuts best at 37 °C in its own unique buffer.
- MspI and its isoschizomer HpaII exhibit different methylation sensitivities
- Source: Moraxella sp.
Storage buffer: 10 mM Tris-HCl (pH 7,4), 50 mM NaCl, 0,1 mM EDTA, 1 mM DTT, 0,5 mg/ml BSA and 50% glycerol.
Fornecimento: Kit includes (1×2000) (1×10 000) U MspI, 1×1 ml buffer B 10X, 1×0,25 ml MULTI-CORE™ 10X buffer, 1×150 µl bovine serum albumin (acetylated).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...