Print…
Caspase-Glo® 3/7 assay kits
Fornecedor: Promega
G8091 G8092 G8090 G8093
PROMG8091EA 542 EUR
PROMG8091 PROMG8092 PROMG8090 PROMG8093
Caspase-Glo® 3/7 assay kits
Ensaios Ensaios celulares
The Caspase-Glo® 3/7 assay is a homogeneous luminescent assay that measures caspase-3/7 activities. The assay provides a proluminescent caspase-3/7 DEVD-aminoluciferin substrate and a proprietary thermostable luciferase in a reagent optimised for caspase-3/7 activity, luciferase activity and cell lysis. This liberates free aminoluciferin, which is consumed by the luciferase, generating a 'glow-type' luminescent signal. The signal is proportional to caspase-3/7 activity. It is designed for use with multiwell plate formats using either purified enzyme or cells in culture.
  • Add-mix-measure protocol makes the assay easy to automate
  • Low background luminescence results in excellent signal-to-noise ratios, allows assay to be performed in 96- or 384-well formats
  • No sample preparation, manipulation and extended incubation times required, maximum sensitivity can be achieved in as little as 15 minutes to 1 hour
  • Delivers excellent Z´ factor values in cell and purified enzyme models
  • Extended-glow signal allows the plates to be read over a 3 hour period of time for batch processing and no injectors required
Fornecimento: The complete assay kit includes Caspase-Glo® 3/7 buffer and Caspase-Glo® 3/7 substrate.