Print…
Gel extractor, x-tracta™
Fornecedor: Promega
A2122 A2121
PROMA2122EA 99.8 EUR
PROMA2122 PROMA2121
Gel extractor, x-tracta™
Puller and Inserter Tools
The x-tracta™ gel extractor tool provides a convenient, safe method for removal of agarose gel fragments for further processing. The device removes a gel piece (3,3×8,4 mm) from agarose gels for easy transfer into a microcentrifuge tube for processing. It eliminates the need for razor blades or scalpels, and its single-use design eliminates the possibility for sample to sample cross contamination.
The x-tracta™ works best on 0,6 to 2% analytical grade agarose gels. It does not work effectively on low melting point agarose gels due to the gel consistency.