DNA purification systems, ReliaPrep™ FFPE gDNA Miniprep System
Fornecedor: Promega
The ReliaPrep™ FFPE gDNA Miniprep System provides a complete, all inclusive method for purifying quality genomic DNA from formalin fixed paraffin-embedded tissue without using hazardous solvents or overnight digestion. The optimised lysis and binding conditions reverse modifications introduced by the fixation process, results in intact, amplifiable gDNA.
- Deparaffinisation step occurs without the use harsh organic solvents
- Saves time – purify gDNA from FFPE tissue in less than two and one-half hours with minimal hands-on time
- No overnight digestion required
- Easy to use – minimal preparation time
Informações de encomenda: Kit components can be ordered separately.
Fornecimento: The complete kit includes ReliaPrep™ FFPE binding columns, collection tubes, proteinase K (PK) solution, RNase A solution, elution buffer, lysis buffer (LBA), BL buffer, wash solution, blue dye, and mineral oil.