Print…
Primers for cDNA synthesis
Fornecedor: Promega
C1101 C1181
PROMC11010020EA 119 EUR
PROMC11010020 PROMC11810020
Primers for cDNA synthesis
Reagentes de ácidos nucleicos Primers and Probes
Oligo (dT)₁₅ primer is suitable for first-strand cDNA synthesis with a reverse transcriptase. The primer hybridises to the poly(A) tail of mRNA. Random hexamers are primers used for first-strand cDNA synthesis and cloning.