RNA Purification kits, Maxwell® 16 LEV simplyRNA
Fornecedor: Promega
The Maxwell® 16 LEV simplyRNA kits are for use with the Maxwell® 16 instrument configured with the low elution volume (LEV) high strength magnetic rod and plunger bar adaptor. This RNA purification procedure is a simple method with minimal lysate handling before automated purification on the Maxwell® 16 instrument. The low elution volume is used to generate concentrated high quality RNA suitable for use in downstream applications, such as quantitative RT-PCR.
- Process up to 1 to 16 samples in an hour
Informações de encomenda: The components in the kit can be ordered separately.
Fornecimento: The kit includes Maxwell® 16 high strength LEV magnetic rod and plunger bar adaptor, LEV plungers and LEV elution tubes.