Gel and PCR clean-up system, Wizard® SV
Fornecedor: Promega
The Wizard® SV Gel and PCR Clean-up System is designed to extract and purify DNA fragments of 100 bp to 10 kb from standard or low-melting agarose gels or to purify products directly from PCR and other common reactions, such as restriction digests. The purified DNA can be used for automated fluorescent DNA sequencing, cloning, labeling, restriction enzyme digestion or in vitro transcription/translation without further manipulation.
- Purify DNA fragments or PCR products in as little as 15 minutes
- Up to 95% recovery is achieved depending upon the DNA fragment size
- Purified DNA routinely achieves 700 bases with >98% accuracy in automated fluorescent sequencing
Informações de encomenda: The components in kit can be ordered separately.
Fornecimento: The complete kit includes Membrane Binding Solution and vacuum adapters.