Print…
Plasmid miniprep system, PureYield™
Fornecedor: Promega
A1222 A6812 A1223
PROMA1222EA 570 EUR
PROMA1222 PROMA6812 PROMA1223
Plasmid miniprep system, PureYield™
Reagentes de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
The PureYield™ plasmid Miniprep system is designed to rapidly isolate highly pure plasmid DNA. The system provides a rapid method for purification of up to 15 μg of plasmid DNA from 600 μl to 3 ml of bacteria culture.
  • Improved productivity: rapid protocol purifies plasmid DNA in 10 minutes
  • High purity and concentration of plasmid DNA gives proven performance in transfection, cell free expression and other molecular biology applications
  • Confidence in results: Lysis/neutralisation indicator dye ensures success every time
  • System has been designed for use with centrifugation or vacuum such as Vac-Man® laboratory vacuum manifold
The system incorporates a unique endotoxin removal wash designed to remove substantial amounts of protein, RNA and endotoxin contaminants from purified plasmid DNA. Removal of contaminants improves the robustness of sensitive applications such as eukaryotic transfection, in vitro transcription and coupled in vitro transcription/translation such as TnT® quick coupled transcription/translation system. Purification is achieved without isopropanol precipitation of purified plasmid DNA or extensive centrifugation, providing rapid purification from a single method.
Informações de encomenda: Kit components can be ordered separately.
Fornecimento: The PureYield™ Miniprep system includes PureYield™ mini columns, endotoxin removal wash (ERB), cell lysis buffer (CLC), neutralisation solution (NSC), column wash solution (CWC), elution buffer (EBB) and PureYield™ collection tubes. The start-up kit includes PureYield™ plasmid Miniprep system, welch vacuum pump and Vac-Man® laboratory vacuum manifold (20 sample capacity).