Print…
Master mixes, GoTaq® qPCR master mix
Fornecedor: Promega
A6001 A6002
A6001EA 366 EUR
A6001 A6002
Master mixes, GoTaq® qPCR master mix
Reagentes de ácidos nucleicos qPCR/RT-qPCR Enzymes and Kits
The GoTaq® qPCR Master Mix is optimised for fast and reproducible quantitative PCR assays using a double-stranded DNA binding dye. The master mix is provided as a ready to use, stabilised 2X formulation and contains GoTaq® Hot Start Polymerase, MgCl₂, dNTPs, BRYT Green® dye and a proprietary reaction buffer.
  • Sensitive detection for earlier quantitation of low and high copy number targets over a broad dynamic range
  • Resistant to a wide range of PCR inhibitors
  • Compatible with both fast and standard cycling methods on most Real-Time PCR instruments
  • Exceptional room temperature set-up is easy for automated and high throughput detection
Fornecimento: Supplied with GoTaq® qPCR master mix (2X), a separate tube of CXR reference dye, and nuclease free water.