Print…
DNA purification systems, Wizard® PCR preps
Fornecedor: Promega
A2180 A7170
PROMA2180EA 0 EUR
PROMA2180 PROMA71700050
DNA purification systems, Wizard® PCR preps
Reagentes de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
The Wizard® PCR preps DNA purification system provides a simple, reliable way to purify double-stranded PCR amplified DNA. This system also can be used to purify DNA fragments from agarose gels. The DNA can be eluted in water or TE buffer, free of salts or macromolecular contaminants. Multiple PCR preps may be processed easily at one time with the Vac-Man® naboratory vacuum manifold.
  • Improved productivity – purify PCR products directly from reactions in 15 minutes
  • Flexibility – separate PCR products from other reaction components such as primers and primer-dimers or from gel slices
Informações de encomenda: Kit components can be ordered separately.
Fornecimento: The complete kit includes Wizard® PCR Preps DNA purification resin, Wizard® minicolumns, direct purification buffer and syringe barrels (3 ml).