Print…
DNA purification systems, Wizard® Plus minipreps
Fornecedor: Promega
A7100 A7500 A7510
PROMA71000050EA 161 EUR
PROMA71000050 PROMA75000100 PROMA75100250
DNA purification systems, Wizard® Plus minipreps
Reagentes de ácidos nucleicos Nucleic Acid Purification Kits and Reagents
The resin-based Wizard® Plus Minipreps DNA purification system provides a simple and reliable method for rapid isolation of plasmid DNA. Minipreps may be processed individually or in multiples with the Vac-Man® (20 sample capacity) or Vac-Man® Jr. (2 sample capacity) laboratory vacuum manifold. The purified plasmid can be used directly for automated fluorescent DNA sequencing and restriction digestion without further manipulation and also can be used for in vitro transcription reactions.
  • Purified DNA meets a target of >98% accuracy over 500 bases using pGEM®-3Zf(+) Vector in BigDye® terminator sequencing
  • Entire procedure may be completed in 15 minutes or less
  • Convenient – no phenol extractions or ethanol precipitations required
Informações de encomenda: The components in kit can be ordered separately.
Fornecimento: The complete kit includes Wizard® Minipreps DNA purification resin, Wizard® minicolumns, cell resuspension solution, cell lysis solution, neutralisation solution, syringe barrels (3 ml), column wash solution.