Genomic DNA purification kits, Wizard®
Fornecedor: Promega
The Wizard® Genomic DNA Purification Kit provides a simple, solution-based method for isolation of DNA from white blood cells, tissue culture cells, animal tissue, plant tissue, yeast and gram-positive and gram-negative bacteria. DNA purified with this system is suitable for a variety of applications, including amplification, digestion with restriction endonucleases and membrane hybridisations such as southern and dot/slot blots.
- Rapidly isolate genomic DNA from blood, tissue culture, animal and plant cells, bacteria and yeast in approximately 60 minutes
- Flexibility – genomic DNA purified is suitable for a variety of applications
Fornecimento: Kit contains cell lysis solution, nuclei lysis solution, protein precipitation solution, DNA rehydration solution, and RNase A solution.