Print…
Bottles, Wide Neck, with Screw Cap
Fornecedor: GLASWARENFABRIK KARL HECHT
41324012 41324006 41324030 41324072 41324054 41324018 41324066 41324048 41324036
HECH41324012EA 193 EUR
HECH41324012 HECH41324006 215-2193 201-0495 201-0496 201-0497 201-0498 HECH41324048 HECH41324036
Bottles, Wide Neck, with Screw Cap
Frascos Frascos de Vidro
Clear glass wide mouth bottles, with black PP cap. Ideal for sampling, storing and transporting powders and pastes.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168