Print…
Restriction enzymes, XhoI, FastDigest™, Fermentas
Fornecedor: Thermo Fisher Scientific
FD0694 FD0695
FD0694EA 63.25 EUR
FD0694 FD0695
Restriction enzymes, XhoI, FastDigest™, Fermentas
Reagentes de ácidos nucleicos Restriction Enzymes
FastDigest™ XhoI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest™ and FastDigest™ Green reaction buffers.
  • 100% buffer compatibility with downstream applications
  • Complete digestion in 5 to 15 minutes
  • Direct loading on gels
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Aplicação
Nome da enzima XhoI
Origem
Overhang type 5' overhang
Buffer compatível FastDigest Green Buffer or FastDigest Buffer (colorless)