Print…
HPLC columns, Pinnacle II PAH
Fornecedor: Restek
921950210 921950212 9219412 9219412-700 9219413-700 9219415 9219415-700 921941E 9219422 9219422-700 9219423-700 9219425 9219425-700 921942E 9219452 9219452-700 9219453-700 9219455 9219455-700 921945E 9219462 9219462-700 9219465 9219465-700 9219472 9219472-700 9219473-700 9219475 9219475-700 921947E
REST921950210EA 322 EUR
REST921950210 REST921950212 REST9219412 REST9219412-700 REST9219413-700 REST9219415 REST9219415-700 REST921941E REST9219422 REST9219422-700 REST9219423-700 REST9219425 REST9219425-700 REST921942E REST9219452 REST9219452-700 REST9219453-700 REST9219455 REST9219455-700 REST921945E REST9219462 REST9219462-700 REST9219465 REST9219465-700 REST9219472 REST9219472-700 REST9219473-700 REST9219475 REST9219475-700 REST921947E
HPLC columns, Pinnacle II PAH
Cromatografia, Colunas
Pinnacle II PAH columns are developed specifically for challenging analyses of polycyclic aromatic hydrocarbons (PAHs).
  • Reliable and cost-effective
  • Greater control over quality and reproducibility
  • Stationary phase incorporates a proprietary C18 bonding that enables unique shape selectivity
pH Range: 2,5 to 8