Print…

Anti-CD44 Mouse Monoclonal Antibody (CF594) [clone: HCAM/1097]

Fornecedor: Biotium
BNC941097-500 BNC941097-100
BNC941097-500EA 560 EUR
BNC941097-500 BNC941097-100
Anti-CD44 Mouse Monoclonal Antibody (CF594) [clone: HCAM/1097]
Anticorpos
Recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.

Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: HCAM/1097
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR