Print…
Anti-CD44 Mouse Monoclonal Antibody (Biotin) [clone: HCAM/1097]
Fornecedor: Biotium
BNCB1097-500 BNCB1097-100
BNCB1097-500EA 560 EUR
BNCB1097-500 BNCB1097-100
Anti-CD44 Mouse Monoclonal Antibody (Biotin) [clone: HCAM/1097]
Anticorpos
Recognizes a cell surface glycoprotein of 80-95 kDa (CD44) on lymphocytes, monocytes, and granulocytes. The CD44 family of glycoproteins exists in a number of variant isoforms, the most common being the standard 85-95 kDa or hematopoietic variant (CD44s). Higher molecular weight isoforms are described in epithelial cells (CD44v), which are believed to function in intercellular adhesion and stromal binding. CD44 immunostaining is commonly used for the discrimination of urothelial transitional cell carcinoma in-situ from non-neoplastic changes in the urothelium.
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: HCAM/1097
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse