Print…
Anti-UCHL1 Rabbit Polyclonal Antibody
Fornecedor: Novus Biologicals
NB300-675 NB300-675SS
NOVUNB300-675EA 626 EUR
NOVUNB300-675 NOVUNB300-675SS
Anti-UCHL1 Rabbit Polyclonal Antibody
Anticorpos
Rabbit Polyclonal UCHL1 Antibody. Tested Applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse, Rat, Bovine, Equine, Guinea Pig, Primate, Porcine.
Recommended Dilutions: Immunohistochemistry (Parafin): 2.5-5.0 ?g/ml
Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human