Print…

Anti-TGFA Mouse Monoclonal Antibody (CF640R) [clone: P/T1]

Fornecedor: Biotium
BNC400787-500 BNC400787-100
BNC400787-500EA 560 EUR
BNC400787-500 BNC400787-100
Anti-TGFA Mouse Monoclonal Antibody (CF640R) [clone: P/T1]
Anticorpos
This antibody reacts with the TGF alpha and shows no cross-reaction with EGF and the neuropeptide synenkephalin. The staining with this MAb is completely blocked by the peptide used for raising this antibody. TGF alpha (aa50) is a growth factor with 33% homology to EGF, binds to EGFR, activates tyrosine phosphorylation of the receptor, and stimulates cell proliferation. It plays a role in tumor initiation by inducing the reversible transformed phenotype.

Type: Primary
Antigen: TGFA
Clonality: Monoclonal
Clone: P/T1
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rabbit, Zebrafish
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR