Print…

Anti-IGHG Mouse Monoclonal Antibody (Purified) [clone: ICO-97]

Fornecedor: Biotium
BNUB0758-500 BNUB0758-100
BNUB0758-500EA 511 EUR
BNUB0758-500 BNUB0758-100
Anti-IGHG Mouse Monoclonal Antibody (Purified) [clone: ICO-97]
Anticorpos
Recognizes a protein of 75 kDa, identified as γ heavy chain of human immunoglobulins. It does not cross-react with α (IgA), μ (IgM), ε (IgE), or δ (IgD), heavy chains, T-cells, monocytes, granulocytes, or erythrocytes. This MAb is useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. The most common feature of these malignancies is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant.

Type: Primary
Antigen: IGHG
Clonality: Monoclonal
Clone: ICO-97
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR