Print…
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF555) [clone: 113-1]
Fornecedor: Biotium
BNC550739-500 BNC550739-100
BNC550739-500EA 560 EUR
BNC550739-500 BNC550739-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF555) [clone: 113-1]
Anticorpos
This MAb recognizes a 60 kDa antigen associated with the mitochondria in human cells. It can be used to stain mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignant cells. This antibody is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells, including neurons and embryonic stem cells. Immunostaining pattern with anti-mitochondrial MAb has been reported as a useful discriminatory adjunct in the complex differential diagnosis of granular renal cell tumors. Reportedly, this MAb facilitates the classification of salivary tumors.
Type: Primary
Antigen: Mitochondrial Marker
Clonality: Monoclonal
Clone: 113-1
Conjugation: CF555
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human