Print…

Anti-KRT10 Mouse Monoclonal Antibody (CF488A) [clone: DE-K13]

Fornecedor: Biotium
BNC880696-500 BNC880696-100
BNC880696-500EA 560 EUR
BNC880696-500 BNC880696-100
Anti-KRT10 Mouse Monoclonal Antibody (CF488A) [clone: DE-K13]
Anticorpos
This antibody recognizes cytokeratin 10 (56.5 kDa) and cytokeratin 13 (53 kDa) in Western blotting. It recognizes only cytokeratin 13 in formalin-fixed, paraffin-embedded tissue sections. It does not react with cytokeratin 10 positive, cytokeratin 13 negative epithelia such as epidermis. However, on frozen sections this MAb serves as differentiation-related marker of all stratified epithelia; it stains all suprabasal cells in both cornifying and non-cornifying stratified epithelia and more differentiated cells of squamous carcinomas.

Type: Primary
Antigen: KRT10
Clonality: Monoclonal
Clone: DE-K13
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cat, Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR