Print…

Anti-CD68 Mouse Monoclonal Antibody (CF488A) [clone: C68/684]

Fornecedor: Biotium
BNC880684-500 BNC880684-100
BNC880684-500EA 560 EUR
BNC880684-500 BNC880684-100
Anti-CD68 Mouse Monoclonal Antibody (CF488A) [clone: C68/684]
Anticorpos
This antibody recognizes a glycoprotein of 110 kDa, which is identified as CD68. It is important for identifying macrophages in tissue sections. It stains macrophages in a wide variety of human tissues, including Kupffer cells and macrophages in the red pulp of the spleen, in lamina propria of the gut, in lung alveoli, and in bone marrow. It reacts with myeloid precursors and peripheral blood granulocytes. It also reacts with plasmacytoid T cells, which are supposed to be of monocyte/macrophage origin. It shows strong granular cytoplasmic staining of chronic and acute myeloid leukemia and also reacts with rare cases of true histiocytic neoplasia. Lymphomas are negative or show few granules.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: Clone C68/684
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR