Print…
Anti-CD36 Mouse Monoclonal Antibody (CF594) [clone: 1E8.]
Fornecedor: Biotium
BNC940314-500 BNC940314-100
BNC940314-500EA 560 EUR
BNC940314-500 BNC940314-100
Anti-CD36 Mouse Monoclonal Antibody (CF594) [clone: 1E8.]
Anticorpos
Recognizes a protein of 80 kDa-90 kDa, identified as CD36. It is expressed on platelets, monocytes and macrophages, microvascular endothelial cells, erythrocyte precursors, mammary epithelial cells, and some macrophage derived dendritic cells. CD36 acts as a receptor for thrombospondin (TSP), collagen types I, IV and V, P. falciparum malaria-infected erythrocytes, and sickle erythrocytes. It also functions as a scavenger receptor, mediating macrophage uptake of oxidized low-density lipoprotein (LDL) and recognition of apoptotic polymorphonuclear leukocytes (PMN). CD36 plays a role in platelet aggregation, macrophage foam cell development, inflammation, and the tissue ischemia observed in sickle cell disease and cerebral malaria. Note that 1-4% of Japanese and East Asia population lack CD36.
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: 1E8.
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human