Print…
Anti-Testosterone Mouse Monoclonal Antibody (Biotin) [clone: 4E1G2]
Fornecedor: Biotium
BNCB0250-500 BNCB0250-100
BNCB0250-500EA 560 EUR
BNCB0250-500 BNCB0250-100
Anti-Testosterone Mouse Monoclonal Antibody (Biotin) [clone: 4E1G2]
Anticorpos
This MAb is highly specific to testosterone. Its affinity constant for testosterone is ~1010M-1. In competitive binding immunoassay, it reacts with testosterone 100%, 11-beta-hydroxy testosterone 3.3%, 17-alpha-methyl testosterone
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: 4E1G2
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: All species