Print…

Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF594) [clone: BM-1]

Fornecedor: Biotium
BNC940061-500 BNC940061-100
BTIUBNC940061-500EA 560 EUR
BTIUBNC940061-500 BTIUBNC940061-100
Anti-Granulocyte Marker Mouse Monoclonal Antibody (CF594) [clone: BM-1]
Anticorpos
Recognizes 183 kDa protein with DNA-binding characteristics, which is identified as a myeloid specific antigen. It reacts with myeloid precursor cells and granulocytes in bone marrow. Its antigen appears to be restricted to M2 and M3 acute myelogenous leukemia (AML) subtypes. Markers of myeloid cells are useful in the identification of different levels of cellular differentiation. This MAb reacts with early precursor and mature forms of human myeloid cells. It is useful in the identification of myelogenous leukemias, distinguishing granulocytic sarcomas from lymphoid malignancies and also in the study of differentiation and transformation of human myeloid cells. The biological function of this antigen is not clear, although it has been proposed that it may play a role in the differentiation of myeloid cells.

Type: Primary
Antigen: Granulocyte Marker
Clonality: Monoclonal
Clone: BM-1
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR