Print…

Anti-ECM1 Mouse Monoclonal Antibody (CF488A) [clone: SC05]

Fornecedor: Biotium
BNC880050-500 BNC880050-100
BNC880050-500EA 560 EUR
BNC880050-500 BNC880050-100
Anti-ECM1 Mouse Monoclonal Antibody (CF488A) [clone: SC05]
Anticorpos
This MAb reacts with a reduction-resistant epitope present in both free and SIgA bound Secretory Component. It does not react with the cell lines lacking secretory component. The antibody is useful for studying the distribution and level of both free and bound secretory component. Secretory component is differentially expressed in epithelium, and the antibody is a popular marker for identifying subpopulations of epithelial cells and epithelial differentiation. The Secretory component antibody is a useful research tool for studying mucosal immunity, inflammation, remodeling, differentiation and tumorigenesis, all processes associated with differential secretory component expression.

Type: Primary
Antigen: ECM1
Clonality: Monoclonal
Clone: SC05
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR