Print…

Anti-Plasma Cell Marker Mouse Monoclonal Antibody (CF405S) [clone: 7B18]

Fornecedor: Biotium
BNC040047-500 BNC040047-100
BNC040047-500EA 560 EUR
BNC040047-500 BNC040047-100
Anti-Plasma Cell Marker Mouse Monoclonal Antibody (CF405S) [clone: 7B18]
Anticorpos
This antibody recognizes an intra-cytoplasmic antigen, which shows a very high degree of specificity for plasma cells. This antigen is present in normal as well as neoplastic plasma cells. Plasma cells, which are large lymphocytes derived from an antigen-specific B cell, secrete antibodies and are responsible for humoral immunity. Plasma cells differentiate from B cells upon stimulation by CD4 lymphocytes. The B cell acts as an antigen-presenting cell (APC), consuming an offending pathogen, which is taken up by the B cell by phagocytosis and broken down within proteosomes. Plasma cells contain basophilic cytoplasm; their nucleus contains heterochromatin organized in a characteristic cartwheel arrangement. This MAb superbly recognizes normal and neoplastic plasma cells in routine formalin-fixed, paraffin-embedded tissue sections. It is of potential value in identifying myeloma or plasmacytoma in bone marrow or other tissues. It also helps differentiate lympho-plasmacytoid lymphoma from lymphocytic and follicular lymphoma. Note that this MAb is not suitable for staining frozen tissues.

Type: Primary
Antigen: Plasma Cell Marker
Clonality: Monoclonal
Clone: 7B18
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR