Print…
Anti-IGKC Mouse Monoclonal Antibody (CF647) [clone: L1C1]
Fornecedor: Biotium
BNC470018-500 BNC470018-100
BNC470018-500EA 560 EUR
BNC470018-500 BNC470018-100
Anti-IGKC Mouse Monoclonal Antibody (CF647) [clone: L1C1]
Anticorpos
This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of kappa to lambda is 70:30. However, with the occurrence of multiple myeloma or other B-cell malignancies this ratio is disturbed. Antibody to the kappa light chain is reportedly useful in the identification of leukemias, plasmacytomas, and certain non-Hodgkin's lymphomas. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is malignant.
Type: Primary
Antigen: IGKC
Clonality: Monoclonal
Clone: L1C1
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human