Print…
Analytical HPLC columns, Ultra Cyano (USP L10)
Fornecedor: Restek
REST910653EEA 552 EUR
REST910653E REST910657E REST9106365-700 REST910633E REST9106313-700 REST9106525-700 REST9106315-700 REST9106332-700 REST9106535 REST9106535-700 REST9106575 REST9106531 REST9106332 REST9106552-700 REST9106335 REST9106532 REST9106571 REST9106331 REST9106363-700 REST9106572 REST9106333-700 REST9106553-700 REST9106523-700 REST910652E REST910636E REST910656E REST9106555-700 REST9106572-700 REST9106525 REST9106365 REST9106522 REST9106521 REST9106565 REST9106352-700 REST9106362 REST9106513-700 REST9106361 REST9106562 REST9106561 REST9106335-700 REST9106573-700 REST910631E REST910651E REST910635E REST910655E REST9106512-700 REST9106522-700 REST9106315 REST9106512 REST9106515 REST9106362-700 REST9106355 REST9106311 REST9106552 REST9106555 REST9106511 REST9106312 REST9106353-700 REST9106565-700 REST9106351 REST9106562-700 REST9106551 REST9106575-700 REST9106352 REST910650210 REST9106532-700 REST910650212 REST9106533-700 REST9106312-700 REST9106355-700 REST9106515-700 REST9106563-700
Analytical HPLC columns, Ultra Cyano (USP L10)
Cromatografia, Colunas
These analytical HPLC columns with spherical shaped traditional monomeric cyanopropylsilane are well suitable for the analysis of protonated bases and also useful for confirmation of analysis on a C18 or C8 columns.
  • Can be used in normal-phase, reversed-phase and HILIC modes
  • Suited with mobile phase when the pH is between 5 and 7
  • Coulmns with trident integral inlet fittings are also available