Print…
Cloreto de ferro (III) hexahidratado, sólido grumoso
Fornecedor: MP Biomedicals
Perigo
Sinônimos: Iron trichloride hexahydrate, Ferric chloride hexahydrate
0215350080 0215350090 0215350091
ICNA0215350080EA 27.6 EUR
ICNA0215350080 ICNA0215350090 ICNA0215350091
Cloreto de ferro (III) hexahidratado, sólido grumoso
Cloreto de Ferro (III) hexahidratado
Soluble in water, alcohol, ether, acetone; practically insoluble in ethyl acetate.
Ferric Chloride is used in photoengraving, photography, the manufacture of pigments and ink, the chlorination of silver and copper ores, and as a mordant in dyeing and printing textiles. It is also utilized in the synthesis of organic compounds. FeCl3 is used in the Mayer's tannic acid/ferric chloride method for staining tissue and their observation by light microscopy. Studies of iron uptake and of apoptosis in cultured cells have utilized FeCl3 as an iron source.
Soluble in water, alcohol, ether, acetone; practically insoluble in ethyl acetate.
Atenção: Research Use Only (RUO).
Fórmula: FeCl₃·6H₂O
Peso Molecular: 270.3 g/mol
Ponto de Fusão: 37
Número MDL : MFCD00149712
Número CAS: 10025-77-1
UN: 3260
ADR: 8 III
Merck Index: 13,04048
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=808c75376e55f29296d58eb1e5a1e184039ce733&vid=6d820873011bee3ff401441544f7f85599e40626&prid=web&clid=SAPDAM
Encomendar
Identity Test Passes
pH (2% aq soln) 1.5 - 2.5